Assay Details
| Gene Symbol: rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002741.2 | NP_001002741.1 | 4 - 5 | 861 | 65 | |
| XM_005169285.4 | XP_005169342.1 | 4 - 5 | 952 | 65 | ||
| GenBank mRNA | BC076412.1 | - | 4 - 5 | 865 | 65 | |
More Information
Set Membership: |
|

rnpep
Genomic Map