Assay Details
| Gene Symbol: acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003746.1 | NP_001003746.1 | 7 - 8 | 801 | 62 | |
| GenBank mRNA | BC078651.1 | - | 7 - 8 | 801 | 62 | |
More Information
Set Membership: |
|

acat1
Genomic Map