Assay Details
| Gene Symbol: cthl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005400.1 | NP_001005400.1 | 10 - 11 | 1091 | 91 | |
| GenBank mRNA | BC080251.1 | - | 10 - 11 | 1091 | 91 | |
More Information
Set Membership: |
|

cthl
Genomic Map