Assay Details
| Gene Symbol: cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017874.1 | NP_001017874.1 | 2 - 3 | 137 | 59 | |
| GenBank mRNA | BC092973.1 | - | 2 - 3 | 137 | 59 | |
More Information
Set Membership: |
|

cnep1r1
Genomic Map