Assay Details
| Gene Symbol: alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020696.1 | NP_001018532.1 | 5 - 6 | 747 | 78 | |
| XM_005171382.3 | XP_005171439.1 | 2 - 3 | 241 | 78 | ||
| GenBank mRNA | BC095802.1 | - | 5 - 6 | 747 | 78 | |
| BC164529.1 | - | 5 - 6 | 743 | 78 | ||
More Information
Set Membership: |
|

alg3
Genomic Map