Assay Details
| Gene Symbol: trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037392.1 | NP_001032469.1 | 9 - 10 | 1218 | 68 | |
| GenBank mRNA | BC109462.1 | - | 9 - 10 | 1218 | 68 | |
More Information
Set Membership: |
|

trmt6
Genomic Map