Assay Details
| Gene Symbol: dync1i2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076667.1 | NP_001070135.1 | 6 - 7 | 591 | 110 | |
| XM_017357723.2 | XP_017213212.2 | 8 - 9 | 847 | 110 | ||
| XM_021478769.1 | XP_021334444.1 | 7 - 8 | 793 | 110 | ||
| GenBank mRNA | BC124315.1 | - | 6 - 7 | 591 | 110 | |
| BC165596.1 | - | 3 - 4 | 305 | 110 | ||
More Information
Set Membership: |
|

dync1i2a
Genomic Map