Assay Details
| Gene Symbol: atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089352.1 | NP_001082821.1 | 9 - 10 | 830 | 70 | |
| XM_005165923.3 | XP_005165980.1 | 9 - 10 | 929 | 70 | ||
| XM_017356471.2 | XP_017211960.1 | 9 - 10 | 934 | 70 | ||
| GenBank mRNA | BC076463.1 | - | 8 - 9 | 814 | 70 | |
| BC134886.1 | - | 9 - 10 | 830 | 70 | ||
More Information
Set Membership: |
|

atg4b
Genomic Map