Assay Details
| Gene Symbol: LOC556326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099446.1 | NP_001092916.1 | 11 - 12 | 1293 | 126 | |
| GenBank mRNA | CU458952.1 | - | 11 - 12 | 1293 | 126 | |
| Gene Symbol: il11ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113499.1 | NP_001106971.1 | 11 - 12 | 1249 | 126 | |
| XM_009305183.3 | XP_009303458.1 | 11 - 12 | 1323 | 126 | ||
More Information
Set Membership: |
|

LOC556326
Genomic Map