Assay Details
| Gene Symbol: gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102475.2 | NP_001095945.1 | 4 - 5 | 471 | 57 | |
| NM_200489.3 | NP_956783.3 | 5 - 6 | 492 | 57 | ||
| GenBank mRNA | AB290319.1 | - | 4 - 5 | 336 | 57 | |
| AB290320.1 | - | 5 - 6 | 357 | 57 | ||
| CK686273.1 | - | 4 - 5 | 475 | 57 | ||
| EH445452.1 | - | 4 - 5 | 417 | 57 | ||
| EH447791.1 | - | 5 - 6 | 449 | 57 | ||
More Information
Set Membership: |
|

gmds
Genomic Map