Assay Details
| Gene Symbol: wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130950.1 | NP_571025.1 | 1 - 2 | 245 | 75 | |
| GenBank mRNA | BC056277.1 | - | 1 - 2 | 260 | 75 | |
| U51266.1 | - | 1 - 2 | 245 | 75 | ||
More Information
Set Membership: |
|

wnt2
Genomic Map