Assay Details
| Gene Symbol: wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130950.1 | NP_571025.1 | 2 - 3 | 471 | 76 | |
| GenBank mRNA | BC056277.1 | - | 2 - 3 | 486 | 76 | |
| U51266.1 | - | 2 - 3 | 471 | 76 | ||
More Information
Set Membership: |
|

wnt2
Genomic Map