Assay Details
| Gene Symbol: etv5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309209.1 | NP_001296138.1 | 5 - 6 | 445 | 96 | |
| NM_131205.2 | NP_571280.2 | 5 - 6 | 445 | 96 | ||
| XM_005165860.4 | XP_005165917.1 | 5 - 6 | 465 | 96 | ||
| XM_005165861.4 | XP_005165918.1 | 5 - 6 | 465 | 96 | ||
| GenBank mRNA | AF168007.1 | - | 4 - 5 | 328 | 96 | |
| BC060924.1 | - | 5 - 6 | 426 | 96 | ||
| BC153467.1 | - | 5 - 6 | 445 | 96 | ||
More Information
Set Membership: |
|

etv5b
Genomic Map