Assay Details
| Gene Symbol: etv5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309209.1 | NP_001296138.1 | 7 - 8 | 852 | 61 | |
| NM_131205.2 | NP_571280.2 | 7 - 8 | 852 | 61 | ||
| XM_005165860.4 | XP_005165917.1 | 7 - 8 | 872 | 61 | ||
| XM_005165861.4 | XP_005165918.1 | 7 - 8 | 872 | 61 | ||
| GenBank mRNA | AF168007.1 | - | 6 - 7 | 736 | 61 | |
| BC060924.1 | - | 7 - 8 | 833 | 61 | ||
| BC153467.1 | - | 7 - 8 | 852 | 61 | ||
| CT594467.2 | - | 3 - 4 | 428 | 61 | ||
More Information
Set Membership: |
|

etv5b
Genomic Map