Assay Details
| Gene Symbol: etv5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309209.1 | NP_001296138.1 | 8 - 9 | 1146 | 123 | |
| NM_131205.2 | NP_571280.2 | 8 - 9 | 1146 | 123 | ||
| GenBank mRNA | AF168007.1 | - | 7 - 8 | 1037 | 124 | |
| BC060924.1 | - | 8 - 9 | 1127 | 123 | ||
| BC153467.1 | - | 8 - 9 | 1146 | 123 | ||
| CT594467.2 | - | 4 - 5 | 722 | 123 | ||
More Information
Set Membership: |
|

etv5b
Genomic Map