Assay Details
| Gene Symbol: mespaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131551.2 | NP_571626.1 | 1 - 2 | 601 | 75 | |
| GenBank mRNA | AB037939.1 | - | 1 - 2 | 602 | 75 | |
| AF188833.1 | - | 1 - 2 | 584 | 75 | ||
| BC162110.1 | - | 1 - 2 | 602 | 75 | ||
| BC162125.1 | - | 1 - 2 | 602 | 75 | ||
More Information
Set Membership: |
|

mespaa
Genomic Map