Assay Details
| Gene Symbol: tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153669.2 | NP_705955.2 | 3 - 4 | 378 | 73 | |
| GenBank mRNA | AY029529.1 | - | 3 - 4 | 387 | 73 | |
| BC052143.1 | - | 3 - 4 | 438 | 73 | ||
| BC063238.1 | - | 3 - 4 | 378 | 73 | ||
More Information
Set Membership: |
|

tram1
Genomic Map