Assay Details
| Gene Symbol: wnt2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182876.2 | NP_878296.1 | 3 - 4 | 844 | 80 | |
| GenBank mRNA | AF544026.1 | - | 3 - 4 | 847 | 80 | |
| BC162076.1 | - | 3 - 4 | 697 | 80 | ||
| BC162077.1 | - | 3 - 4 | 697 | 80 | ||
More Information
Set Membership: |
|

wnt2ba
Genomic Map