Assay Details
| Gene Symbol: hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199599.2 | NP_955893.1 | 2 - 3 | 269 | 67 | |
| XM_005155591.4 | XP_005155648.1 | 3 - 4 | 419 | 67 | ||
| XM_005155592.4 | XP_005155649.1 | 3 - 4 | 416 | 67 | ||
| GenBank mRNA | BC059502.1 | - | 2 - 3 | 302 | 67 | |
| BC062838.1 | - | 2 - 3 | 269 | 67 | ||
More Information
Set Membership: |
|

hsdl2
Genomic Map