Assay Details
| Gene Symbol: synj2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199917.1 | NP_956211.1 | 2 - 3 | 306 | 148 | |
| GenBank mRNA | BC045991.1 | - | 2 - 3 | 306 | 148 | |
More Information
Set Membership: |
|

synj2bp
Genomic Map