Assay Details
| Gene Symbol: sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199976.1 | NP_956270.1 | 4 - 5 | 579 | 66 | |
| GenBank mRNA | AY195857.1 | - | 4 - 5 | 578 | 66 | |
| BC060895.1 | - | 4 - 5 | 579 | 66 | ||
More Information
Set Membership: |
|

sod2
Genomic Map