Assay Details
| Gene Symbol: abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200157.1 | NP_956451.1 | 1 - 2 | 749 | 76 | |
| GenBank mRNA | BC045350.1 | - | 1 - 2 | 749 | 76 | |
| BC152232.1 | - | 1 - 2 | 770 | 76 | ||
More Information
Set Membership: |
|

abhd17c
Genomic Map