Assay Details
| Gene Symbol: abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200157.1 | NP_956451.1 | 2 - 3 | 939 | 65 | |
| GenBank mRNA | BC045350.1 | - | 2 - 3 | 939 | 65 | |
More Information
Set Membership: |
|

abhd17c
Genomic Map