Assay Details
| Gene Symbol: mfsd4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200531.1 | NP_956825.1 | 7 - 8 | 1409 | 79 | |
| GenBank mRNA | BC055671.1 | - | 7 - 8 | 1409 | 79 | |
More Information
Set Membership: |
|

mfsd4aa
Genomic Map