Assay Details
| Gene Symbol: shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201196.1 | NP_957490.1 | 3 - 4 | 697 | 76 | |
| XM_021481365.1 | XP_021337040.1 | 3 - 4 | 834 | 76 | ||
| XM_021481366.1 | XP_021337041.1 | 4 - 5 | 747 | 76 | ||
| XM_021481367.1 | XP_021337042.1 | 4 - 5 | 744 | 76 | ||
| GenBank mRNA | BC056324.1 | - | 3 - 4 | 697 | 76 | |
| BC065345.1 | - | 3 - 4 | 697 | 76 | ||
More Information
Set Membership: |
|

shox2
Genomic Map