Assay Details
| Gene Symbol: tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205677.1 | NP_991240.1 | 5 - 6 | 814 | 81 | |
| GenBank mRNA | BC065467.1 | - | 5 - 6 | 814 | 81 | |
More Information
Set Membership: |
|

tmem45b
Genomic Map