Assay Details
| Gene Symbol: cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205700.1 | NP_991263.1 | 3 - 4 | 458 | 124 | |
| GenBank mRNA | BC065947.1 | - | 3 - 4 | 458 | 124 | |
More Information
Set Membership: |
|

cfl2
Genomic Map