Assay Details
| Gene Symbol: tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207100.2 | NP_996983.1 | 739 | 109 | ||
| GenBank mRNA | BC124418.1 | - | 739 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem208| Gene Symbol: tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207100.2 | NP_996983.1 | 739 | 109 | ||
| GenBank mRNA | BC124418.1 | - | 739 | 109 | ||
Set Membership: |
|