Assay Details
| Gene Symbol: ap2m1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291355.1 | NP_001278284.1 | 6 - 7 | 740 | 63 | |
| NM_212577.2 | NP_997742.1 | 6 - 7 | 740 | 63 | ||
| GenBank mRNA | BC047180.1 | - | 6 - 7 | 737 | 63 | |
| BC067560.1 | - | 5 - 6 | 619 | 63 | ||
| BC164210.1 | - | 5 - 6 | 597 | 63 | ||
| EH432280.1 | - | 6 - 7 | 750 | 63 | ||
| EH456153.1 | - | 6 - 7 | 740 | 63 | ||
| EH494715.1 | - | 6 - 7 | 722 | 63 | ||
More Information
Set Membership: |
|

ap2m1a
Genomic Map