Assay Details
| Gene Symbol: ap2m1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291355.1 | NP_001278284.1 | 11 - 12 | 1255 | 69 | |
| NM_212577.2 | NP_997742.1 | 10 - 11 | 1240 | 69 | ||
| GenBank mRNA | BC047180.1 | - | 10 - 11 | 1237 | 69 | |
| BC067560.1 | - | 9 - 10 | 1119 | 69 | ||
| BC164210.1 | - | 9 - 10 | 1097 | 69 | ||
| EB766597.1 | - | 3 - 4 | 321 | 69 | ||
More Information
Set Membership: |
|

ap2m1a
Genomic Map