Assay Details
| Gene Symbol: sntb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002175.1 | NP_001002175.1 | 1 - 2 | 547 | 67 | |
| GenBank mRNA | BC074093.1 | - | 1 - 2 | 547 | 67 | |
| BC165289.1 | - | 1 - 2 | 531 | 67 | ||
More Information
Set Membership: |
|

sntb1
Genomic Map