Assay Details
| Gene Symbol: tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002637.1 | NP_001002637.1 | 2 - 3 | 304 | 64 | |
| GenBank mRNA | BC075937.1 | - | 2 - 3 | 304 | 64 | |
More Information
Set Membership: |
|

tmem53
Genomic Map