Assay Details
| Gene Symbol: wdr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003630.2 | NP_001003630.2 | 7 - 8 | 913 | 91 | |
| XM_005165298.4 | XP_005165355.1 | 7 - 8 | 941 | 91 | ||
| GenBank mRNA | BC077087.1 | - | 7 - 8 | 917 | 91 | |
More Information
Set Membership: |
|

wdr54
Genomic Map