Assay Details
| Gene Symbol: phyhiplb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005602.2 | NP_001005602.2 | 2 - 3 | 493 | 88 | |
| XM_005158004.4 | XP_005158061.1 | 1 - 2 | 185 | 88 | ||
| GenBank mRNA | BC139549.1 | - | 2 - 3 | 493 | 88 | |
More Information
Set Membership: |
|

phyhiplb
Genomic Map