Assay Details
| Gene Symbol: magi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007063.2 | NP_001007064.2 | 11 - 12 | 2026 | 78 | |
| XM_005155829.4 | XP_005155886.1 | 11 - 12 | 2060 | 78 | ||
| XM_005155830.4 | XP_005155887.1 | 11 - 12 | 2060 | 78 | ||
| XM_005155831.4 | XP_005155888.1 | 11 - 12 | 2061 | 78 | ||
| XM_009305951.3 | XP_009304226.1 | 11 - 12 | 2042 | 78 | ||
| XM_009305953.3 | XP_009304228.1 | 10 - 11 | 1925 | 78 | ||
| XM_009305956.3 | XP_009304231.1 | 11 - 12 | 2061 | 78 | ||
| XM_021479602.1 | XP_021335277.1 | 10 - 11 | 2024 | 78 | ||
| GenBank mRNA | AY465352.1 | - | 11 - 12 | 2026 | 78 | |
More Information
Set Membership: |
|

magi1b
Genomic Map