Assay Details
| Gene Symbol: ripor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012499.2 | NP_001012517.1 | 2 - 3 | 229 | 71 | |
| XM_005166267.4 | XP_005166324.1 | 2 - 3 | 417 | 71 | ||
| XM_009302697.3 | XP_009300972.1 | 2 - 3 | 383 | 71 | ||
| GenBank mRNA | BC090162.1 | - | 2 - 3 | 233 | 71 | |
More Information
Set Membership: |
|

ripor3
Genomic Map