Assay Details
| Gene Symbol: med4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017883.1 | NP_001017883.1 | 5 - 6 | 477 | 81 | |
| GenBank mRNA | BC092965.1 | - | 5 - 6 | 477 | 81 | |
| BC164333.1 | - | 5 - 6 | 495 | 81 | ||
More Information
Set Membership: |
|

med4
Genomic Map