Assay Details
| Gene Symbol: slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020474.1 | NP_001018310.1 | 4 - 5 | 814 | 64 | |
| XM_005166394.4 | XP_005166451.1 | 5 - 6 | 1074 | 64 | ||
| GenBank mRNA | AB193556.1 | - | 4 - 5 | 814 | 64 | |
More Information
Set Membership: |
|

slc7a6
Genomic Map