Assay Details
| Gene Symbol: trnau1apb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029966.2 | NP_001025137.2 | 3 - 4 | 340 | 89 | |
| XM_021481362.1 | XP_021337037.1 | 3 - 4 | 361 | 89 | ||
| GenBank mRNA | BC098884.1 | - | 3 - 4 | 340 | 89 | |
| CV489035.1 | - | 3 - 4 | 350 | 89 | ||
More Information
Set Membership: |
|

trnau1apb
Genomic Map