Assay Details
| Gene Symbol: trnau1apb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029966.2 | NP_001025137.2 | 4 - 5 | 397 | 86 | |
| XM_021481362.1 | XP_021337037.1 | 4 - 5 | 418 | 86 | ||
| GenBank mRNA | BC063993.1 | - | 3 - 4 | 833 | 86 | |
| BC098884.1 | - | 4 - 5 | 397 | 86 | ||
| CV489035.1 | - | 4 - 5 | 407 | 86 | ||
More Information
Set Membership: |
|

trnau1apb
Genomic Map