Assay Details
| Gene Symbol: trnau1apb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029966.2 | NP_001025137.2 | 6 - 7 | 648 | 96 | |
| GenBank mRNA | BC063993.1 | - | 6 - 7 | 1133 | 96 | |
| BC098884.1 | - | 6 - 7 | 648 | 96 | ||
| EB769378.1 | - | 1 - 2 | 53 | 96 | ||
More Information
Set Membership: |
|

trnau1apb
Genomic Map