Assay Details
| Gene Symbol: sid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034182.2 | NP_001029354.2 | 1386 | 117 | ||
| GenBank mRNA | AY494978.1 | - | 1146 | 117 | ||
| BC134848.1 | - | 1386 | 117 | |||
| CU458937.1 | - | 1170 | 117 | |||
More Information
Set Membership: |
|

sid4
Genomic Map