Assay Details
| Gene Symbol: rdh14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047190.1 | NP_001040655.1 | 1 - 2 | 376 | 87 | |
| GenBank mRNA | BC095278.1 | - | 1 - 2 | 376 | 87 | |
More Information
Set Membership: |
|

rdh14b
Genomic Map