Assay Details
| Gene Symbol: narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080991.2 | NP_001074460.2 | 10 - 11 | 1214 | 64 | |
| GenBank mRNA | BC131874.1 | - | 10 - 11 | 1121 | 64 | |
More Information
Set Membership: |
|

narfl
Genomic Map