Assay Details
| Gene Symbol: march5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082827.2 | NP_001076296.1 | 5 - 6 | 1023 | 69 | |
| GenBank mRNA | BC134171.1 | - | 5 - 6 | 1024 | 69 | |
More Information
Set Membership: |
|

march5
Genomic Map