Assay Details
| Gene Symbol: fscn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005170877.4 | XP_005170934.1 | 2 - 3 | 983 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fscn1b| Gene Symbol: fscn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005170877.4 | XP_005170934.1 | 2 - 3 | 983 | 102 | |
Set Membership: |
|