Assay Details
| Gene Symbol: gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089386.1 | NP_001082855.1 | 1 - 2 | 220 | 109 | |
| GenBank mRNA | BC139530.1 | - | 1 - 2 | 220 | 109 | |
More Information
Set Membership: |
|

gfer
Genomic Map