Assay Details
| Gene Symbol: tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089537.1 | NP_001083006.1 | 9 - 10 | 1256 | 62 | |
| XM_005157147.4 | XP_005157204.1 | 8 - 9 | 1186 | 62 | ||
| XM_009307730.3 | XP_009306005.1 | 9 - 10 | 1288 | 62 | ||
| GenBank mRNA | BC139553.1 | - | 9 - 10 | 1256 | 62 | |
More Information
Set Membership: |
|

tmco6
Genomic Map