Assay Details
| Gene Symbol: myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131329.3 | NP_571404.1 | 6 - 7 | 594 | 74 | |
| GenBank mRNA | AF114428.1 | - | 4 - 5 | 308 | 74 | |
| AF425743.1 | - | 5 - 6 | 421 | 74 | ||
| AY057074.1 | - | 6 - 7 | 463 | 74 | ||
| BC076232.1 | - | 6 - 7 | 595 | 74 | ||
More Information
Set Membership: |
|

myl7
Genomic Map