Assay Details
| Gene Symbol: sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131756.1 | NP_571831.1 | 8 - 9 | 963 | 74 | |
| GenBank mRNA | AJ009864.1 | - | 8 - 9 | 963 | 74 | |
| BC071295.1 | - | 8 - 9 | 938 | 74 | ||
More Information
Set Membership: |
|

sms
Genomic Map